accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3OOCH3O

CAS RN 613-03-6

IUPAC Name: 2,4-bis(acetyloxy)phenyl acetate
Synonyms: 1,2,4-benzenetriol, triacetate
Molecular Weight (g/mol): 252.22
Molecular Formula: C12H12O6
InChi Key: AESFGSJWSUZRGW-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(OC(C)=O)C(OC(C)=O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)