Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Triacetoxybenzene 97%, Thermo Scientific™

Catalog No. AC157891000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC157891000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC157891000 Supplier Thermo Scientific Chemicals Supplier No. 157891000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-03-6
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.222
InChI Key AESFGSJWSUZRGW-UHFFFAOYSA-N
Synonym 1,2,4-triacetoxybenzene, benzene-1,2,4-triyl triacetate, hydroxyhydroquinone triacetate, 1,2,4-benzenetriol, triacetate, pyrogallol a, 2-hydroxyhydroquinone triacetate, 1,2,4-benzenetriacetate, unii-903t7h7cp1, triacetate d'hydroxyhydroquinone, 2,4-bis acetyloxy phenyl acetate
PubChem CID 69169
IUPAC Name (3,4-diacetyloxyphenyl) acetate
SMILES CC(=O)OC1=CC(=C(C=C1)OC(=O)C)OC(=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 98°C to 100°C
Color Beige
Infrared Spectrum Authentic
Linear Formula (CH3CO2)3C6H3
Quantity 100 g
Beilstein 06, 1089
Formula Weight 252.22
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1, 2, 4-Triacetoxybenzene, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.