accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 613-31-0

IUPAC Name: 9,10-dihydroanthracene
Synonyms: 9,10-dihydroanthracene
Molecular Weight (g/mol): 180.25
Molecular Formula: C14H12
InChi Key: WPDAVTSOEQEGMS-UHFFFAOYSA-N
SMILES: C1C2=CC=CC=C2CC2=CC=CC=C12
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)