Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Dihydroanthracene (Tech.), 90%, Thermo Scientific™

Catalog No. AC148222500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC148222500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC148222500 Supplier Thermo Scientific Chemicals Supplier No. 148222500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-31-0
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
InChI Key WPDAVTSOEQEGMS-UHFFFAOYSA-N
Synonym anthracene, 9,10-dihydro, anthracene, dihydro, 9,10-dihydro-anthracene, unii-z142c238gb, anthracene,10-dihydro, acmc-1bh69, 9,10-dihydroanthracen-9-yl, 9,10-dihydroanthracene
PubChem CID 11940
IUPAC Name 9,10-dihydroanthracene
SMILES C1C2=CC=CC=C2CC3=CC=CC=C31

Specifications

CAS Min % 88.0
CAS Max % 100.0
Density 0.88
Quantity 250 g
Beilstein 05, 641
Formula Weight 180.25
Grade Technical
Chemical Name or Material 9, 10-Dihydroanthracene, tech., 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.