accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH3C

CAS RN 613-45-6

IUPAC Name: 2,4-dimethoxybenzaldehyde
Synonyms: 2,4-dimethoxybenzaldehyd
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: LWRSYTXEQUUTKW-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C(OC)=C1
Boiling Point 
  • (3)
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (1)
  • (3)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (6)
  • (3)
  • (4)
  • (3)
  • (1)
  • (6)
  • (3)
  • (3)
  • (7)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)