Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxybenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. D0626500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0626500G 500 g
D062625G 25 g
D0626100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0626500G Supplier TCI America Supplier No. D0626500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 613-45-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00003311
InChI Key LWRSYTXEQUUTKW-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dimethoxy, 2,4-dimethoxybenzaldehyd, 2,4-dimethoxy-benzaldehyde, 2,4-bis methyloxy benzaldehyde, ameba resin, pubchem8220, acmc-209mro, 2,4dimethoxybenzaldehyde, argopore tm-mb-cho, 2,4-dimethoxybenzaldehye
PubChem CID 69175
IUPAC Name 2,4-dimethoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)C=O)OC

Specifications

Melting Point 71°C
Color White-Yellow
Boiling Point 165°C
Quantity 500 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxybenzaldehyde
EINECSNumber (3)-2665
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.