accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 613-48-9

IUPAC Name: N,N-diethyl-4-methylaniline
Synonyms: N,N-diethyl-4-methylaniline
Molecular Weight (g/mol): 163.26
Molecular Formula: C11H17N
InChi Key: HKJNHYJTVPWVGV-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results