Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyl-p-toluidine 98.0+%, TCI America™
SDP

Catalog No. D053325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D053325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D053325ML Supplier TCI America Supplier No. D053325ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 613-48-9
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00015113
InChI Key HKJNHYJTVPWVGV-UHFFFAOYSA-N
Synonym n,n-diethyl-p-toluidine, benzenamine, n,n-diethyl-4-methyl, nn-diethyl-p-toluidine, 4-diethylamino toluene, p-methyl-n,n-diethylaniline, p-toluidine, n,n-diethyl, n,n-diethyl-p-toluidinium ion, 4-diethylaminotoluene, diethyl 4-methylphenyl amine, diethyl-p-toluidine
PubChem CID 69177
IUPAC Name N,N-diethyl-4-methylaniline
SMILES CCN(CC)C1=CC=C(C=C1)C

Specifications

Color Yellow
Boiling Point 230°C
Quantity 25 mL
UN Number 2810
Formula Weight 163.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-p-toluidine
EINECSNumber (3)-0191
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.