accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 613-62-7

IUPAC Name: 2-(benzyloxy)naphthalene
Synonyms: 2-(benzyloxy)naphthalene
Molecular Weight (g/mol): 234.3
Molecular Formula: C17H14O
InChi Key: WLTCCDHHWYAMCG-UHFFFAOYSA-N
SMILES: C(OC1=CC=C2C=CC=CC2=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results