accessibility menu, dialog, popup

UserName

Viewing profile as user:
ON

CAS RN 613-90-1

IUPAC Name: benzoyl cyanide
Synonyms: benzoyl cyanide α-oxobenzeneacetonitrile benzoyl nitrile α-oxo-α-tolunitrile
Molecular Weight (g/mol): 131.13
Molecular Formula: C8H5NO
InChi Key: GJQBHOAJJGIPRH-UHFFFAOYSA-N
SMILES: O=C(C#N)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)