Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl Cyanide 98.0+%, TCI America™
SDP

Catalog No. B083525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B083525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B083525G Supplier TCI America Supplier No. B083525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 613-90-1
Molecular Formula C8H5NO
Molecular Weight (g/mol) 131.134
MDL Number MFCD00001837
InChI Key GJQBHOAJJGIPRH-UHFFFAOYSA-N
Synonym phenylglyoxylonitrile, benzoyl nitrile, benzoylcyanide, oxo phenyl acetonitrile, alpha-oxobenzeneacetonitrile, benzeneacetonitrile, .alpha.-oxo, glyoxylonitrile, phenyl, alpha-oxo-alpha-tolunitrile, 2-oxo-2-phenylethanenitrile, alpha-tolunitrile, alpha-oxo
PubChem CID 11953
ChEBI CHEBI:51853
IUPAC Name benzoyl cyanide
SMILES C1=CC=C(C=C1)C(=O)C#N

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 131.13
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Benzoyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.