accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61315-61-5

IUPAC Name: 5-{[amino(4-methylbenzenesulfonamido)methylidene]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms: 5-{[amino(4-methylbenzenesulfonamido)methylidene]amino}-2-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol): 428.5
Molecular Formula: C18H28N4O6S
InChi Key: WBIIPXYJAMICNU-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC(N)=NCCCC(NC(=O)OC(C)(C)C)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)