Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boc-D-Arg Tos -OH, Advanced ChemTech

Catalog No. 01340972
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-972 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-340-972 Supplier Advanced Chem Tech Supplier No. BR31255G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 61315-61-5 | C18H28N4O6S | 428.50 g/mol

Chemical Identifiers

CAS 61315-61-5
Molecular Formula C18H28N4O6S
Molecular Weight (g/mol) 428.50
InChI Key WBIIPXYJAMICNU-UHFFFAOYNA-N
IUPAC Name 5-{[amino(4-methylbenzenesulfonamido)methylidene]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(N)=NCCCC(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Color White to Off-White
Quantity 5 g
Grade Biochemical
Chemical Name or Material Boc-D-Arg Tos -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.