CAS RN 6132-04-3
Trisodium citrate dihydrate, 99%
CAS: 6132-04-3 Molecular Formula: C6H9Na3O9 Molecular Weight (g/mol): 294.10 MDL Number: MFCD00150031 InChI Key: NLJMYIDDQXHKNR-UHFFFAOYSA-K Synonym: trisodium citrate dihydrate,sodium citrate dihydrate,sodium citrate tribasic dihydrate,natriumcitrat-dihydrat,sodium citrate hydrate,1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate,citronensaeure-tri-na-salz-dihydrat,citric acid trisodium salt dihydrate,citric acid, trisodium salt dihydrate,trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate PubChem CID: 71474 ChEBI: CHEBI:32142 IUPAC Name: trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
Sodium Citrate Dihydrate, ACS Reagent Grade, Ricca Chemical
CAS: 6132-04-3 Molecular Formula: C6H9Na3O9 Molecular Weight (g/mol): 294.10 InChI Key: NLJMYIDDQXHKNR-UHFFFAOYSA-K Synonym: trisodium citrate dihydrate,sodium citrate dihydrate,sodium citrate tribasic dihydrate,natriumcitrat-dihydrat,sodium citrate hydrate,1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate,citronensaeure-tri-na-salz-dihydrat,citric acid trisodium salt dihydrate,citric acid, trisodium salt dihydrate,trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate PubChem CID: 71474 ChEBI: CHEBI:32142 IUPAC Name: trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
Sodium Citrate Dihydrate (Small, Colorless Granules), Fisher BioReagents™
CAS: 6132-04-3 Molecular Formula: C6H9Na3O9 Molecular Weight (g/mol): 294.10 InChI Key: NLJMYIDDQXHKNR-UHFFFAOYSA-K Synonym: trisodium citrate dihydrate,sodium citrate dihydrate,sodium citrate tribasic dihydrate,natriumcitrat-dihydrat,sodium citrate hydrate,1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate,citronensaeure-tri-na-salz-dihydrat,citric acid trisodium salt dihydrate,citric acid, trisodium salt dihydrate,trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate PubChem CID: 71474 ChEBI: CHEBI:32142 IUPAC Name: trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
Sodium Citrate Dihydrate (Granular/Certified), Fisher Chemical™
Molecular Formula: C6H9Na3O9 Molecular Weight (g/mol): 294.10 MDL Number: MFCD00150031 InChI Key: NLJMYIDDQXHKNR-UHFFFAOYSA-K Synonym: trisodium citrate dihydrate,sodium citrate dihydrate,sodium citrate tribasic dihydrate,natriumcitrat-dihydrat,sodium citrate hydrate,1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate,citronensaeure-tri-na-salz-dihydrat,citric acid trisodium salt dihydrate,citric acid, trisodium salt dihydrate,trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate PubChem CID: 71474 ChEBI: CHEBI:32142 IUPAC Name: trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
Carbon Dioxide ISA, Fisherbrand
CAS: 6132-04-3 Molecular Formula: C6H9Na3O9 Molecular Weight (g/mol): 294.10 InChI Key: NLJMYIDDQXHKNR-UHFFFAOYSA-K PubChem CID: 71474 ChEBI: CHEBI:32142 IUPAC Name: trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
Sodium Citrate, 3.8% (w/v), Ricca Chemical
CAS: 6132-04-3 Molecular Formula: C6H9Na3O9 Molecular Weight (g/mol): 294.10 InChI Key: NLJMYIDDQXHKNR-UHFFFAOYSA-K Synonym: trisodium citrate dihydrate,sodium citrate dihydrate,sodium citrate tribasic dihydrate,natriumcitrat-dihydrat,sodium citrate hydrate,1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate,citronensaeure-tri-na-salz-dihydrat,citric acid trisodium salt dihydrate,citric acid, trisodium salt dihydrate,trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate PubChem CID: 71474 ChEBI: CHEBI:32142 IUPAC Name: trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
Corning™ Molecular Biology Reagents
Used in molecular biology, nucleic acid purification applications
Sodium Citrate, Dihydrate, Granular, USP, 99-100.5%, Spectrum™ Chemical
CAS: 6132-04-3 Molecular Formula: C6H9Na3O9 Molecular Weight (g/mol): 294.10 InChI Key: NLJMYIDDQXHKNR-UHFFFAOYSA-K IUPAC Name: trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate SMILES: O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Cupric Citrate TS, Alkaline, (U.S.P. Test Solution), Spectrum™ Chemical
CAS: 6132-04-3 Molecular Formula: C6H9Na3O9 Molecular Weight (g/mol): 294.10 InChI Key: NLJMYIDDQXHKNR-UHFFFAOYSA-K IUPAC Name: trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate SMILES: O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Sodium Citrate, Dihydrate, Granular, FCC, 99-100.5%, Spectrum™ Chemical
CAS: 6132-04-3 Molecular Formula: C6H9Na3O9 Molecular Weight (g/mol): 294.10 InChI Key: NLJMYIDDQXHKNR-UHFFFAOYSA-K IUPAC Name: trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate SMILES: O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Sodium Citrate, Dihydrate, Crystal, ACS, 99%, Spectrum™ Chemical
CAS: 6132-04-3
Sodium Citrate, 10% (w/w) (10 g + 90 mL Water), Ricca Chemical
CAS: 6132-04-3 Molecular Formula: C6H9Na3O9 Molecular Weight (g/mol): 294.10 InChI Key: NLJMYIDDQXHKNR-UHFFFAOYSA-K Synonym: trisodium citrate dihydrate,sodium citrate dihydrate,sodium citrate tribasic dihydrate,natriumcitrat-dihydrat,sodium citrate hydrate,1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate,citronensaeure-tri-na-salz-dihydrat,citric acid trisodium salt dihydrate,citric acid, trisodium salt dihydrate,trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate PubChem CID: 71474 ChEBI: CHEBI:32142 IUPAC Name: trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
Medisca Sodium Citrate USP/EP (Dihydrate)
Sodium Citrate USP / EP is a white or almost white, crystalline powder or white or almost white, granular crystals. It is slightly deliquescent in moist air.