Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate, Dihydrate, Granular, FCC, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606459 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-459 45 kg Poly Bottle
18-606-454 500 g Poly Bottle
18-606-456 2.5 kg Poly Bottle
18-606-455 6 x 500 g Poly Bottle
18-606-457 4 x 2.5 kg Poly Bottle
18-606-458 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-606-459 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S125645KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1256, 6132-04-3, C6H5Na3O7·2H2O

Spectrum™ Chemical Sodium Citrate, Dihydrate, Granular, FCC is used as a flavorant and preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6132-04-3
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
IUPAC Name trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
SMILES O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 45 kg
Percent Purity 99 to 100.5%
Grade FCC
Packaging Poly Bottle
Chemical Name or Material Sodium Citrate, Dihydrate, Granular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.