accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61323-24-8

CH3CH3SH3CN

CAS RN 61323-24-8

IUPAC Name: 4-methyl-2-(2-methylpropyl)-1,3-thiazole
Synonyms: 4-methyl-2-(2-methylpropyl)-1,3-thiazole
Molecular Weight (g/mol): 155.26
Molecular Formula: C8H13NS
InChi Key: JBUCYVMFLWLDIO-UHFFFAOYSA-N
SMILES: CC(C)CC1=NC(C)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results