Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isobutyl-4-methylthiazole 96.0+%, TCI America™
SDP

Catalog No. I04425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04425ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04425ML Supplier TCI America Supplier No. I04425ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61323-24-8
Molecular Formula C8H13NS
Molecular Weight (g/mol) 155.259
MDL Number MFCD00053138
InChI Key JBUCYVMFLWLDIO-UHFFFAOYSA-N
PubChem CID 574813
IUPAC Name 4-methyl-2-(2-methylpropyl)-1,3-thiazole
SMILES CC1=CSC(=N1)CC(C)C

Specifications

Boiling Point 189°C
Quantity 5 mL
Formula Weight 155.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isobutyl-4-methylthiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.