accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61341-86-4

IUPAC Name: 2,3-dihydro-1H-inden-1-amine
Synonyms: indanamin
Molecular Weight (g/mol): 133.19
Molecular Formula: C9H11N
InChi Key: XJEVHMGJSYVQBQ-UHFFFAOYNA-N
SMILES: NC1CCC2=CC=CC=C12
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)