Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-1-Aminoindan

Catalog No. 445355250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-535-5250MG 250 mg
44-535-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 44-535-5250MG Supplier Sigma Aldrich Supplier No. 445355250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-1-Aminoindan

Specifications

CAS 61341-86-4
Density 1.038 g/mL at 25°C
Boiling Point 96°C to 97°C (8 mmHg, lit.)
Molecular Formula C9H11N
Refractive Index n20/D 1.562
Linear Formula C9H11N
MDL Number MFCD00216670
Quantity 250 mg
Synonym (S)-(+)-1-Indanamine
Molecular Weight (g/mol) 133.19 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.