accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61348-28-5

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-4-carbamoylbutanoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-4-carbamoylbutanoate
Molecular Weight (g/mol): 245.26
Molecular Formula: C10H17N2O5
InChi Key: VVNYDCGZZSTUBC-ZCFIWIBFSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CCC(N)=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)