Learn More
N(alpha)-Boc-D-glutamine, 98+%, Thermo Scientific Chemicals
Description
N2-Boc-D-glutamine is an N-Boc-protected form of D-Glutamine. D-Glutamine is an unnatural isomer of L-Glutamine that is present in human plasma an is a source of liberated ammonia. D-Glutamine can be synthesized by enzymatic means or can be found in cheeses, wine and vinegars as well.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N2-Boc-D-glutamine is an N-Boc-protected form of D-Glutamine. D-Glutamine is an unnatural isomer of L-Glutamine that is present in human plasma an is a source of liberated ammonia. D-Glutamine can be synthesized by enzymatic means or can be found in cheeses, wine and vinegars as well.
Solubility
Soluble, 1mmol in 2mL DMF. Soluble in chloroform and methanol.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications
Specifications
| Melting Point | 117°C to 119°C |
| Beilstein | 1981311 |
| Solubility Information | Soluble,1mmol in 2mL DMF. Soluble in chloroform and methanol. |
| Quantity | 1 g |
| Formula Weight | 246.26 |
| Optical Rotation | +3.5° (c=2 in Ethanol) |
| Percent Purity | ≥98% |
| Chemical Name or Material | N(alpha)-Boc-D-glutamine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.