accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 613660-87-0

IUPAC Name: (4-sulfamoylphenyl)boronic acid
Synonyms: 4-sulfamoylphenylboronic acid
Molecular Weight (g/mol): 201
Molecular Formula: C6H8BNO4S
InChi Key: AKSXQPCIAOIJGP-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results