Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Sulfamoylbenzeneboronic acid, 97%, Thermo Scientific™

Catalog No. AAH52928MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52928MD 250 mg
AAH5292803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52928MD Supplier Thermo Scientific Chemicals Supplier No. H52928MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Sulfamoylbenzeneboronic acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613660-87-0
Molecular Formula C6H8BNO4S
Molecular Weight (g/mol) 201.003
MDL Number MFCD06659839
InChI Key AKSXQPCIAOIJGP-UHFFFAOYSA-N
Synonym 4-aminosulfonylphenyl boronic acid, 4-aminosulfonyl phenylboronic acid, 4-sulfamoylphenyl boronic acid, 4-boronobenzenesulfonamide, 4-aminosulphonyl benzeneboronic acid, 4-aminosulfonyl benzeneboronic acid, 4-sulphamoylbenzeneboronic acid, 4-sulfamoylbenzeneboronic acid, 4-boronobenzenesulphonamide, boronic acid, 4-aminosulfonyl phenyl
PubChem CID 22391977
IUPAC Name (4-sulfamoylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)S(=O)(=O)N)(O)O

Specifications

Melting Point 250°C to 256°C
Quantity 250 mg
Formula Weight 201
Percent Purity 97%
Chemical Name or Material 4-Sulfamoylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.