accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClNbHClCCH3CCCCH3CC

CAS RN 61374-51-4

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 320.04
Molecular Formula: C12H12Cl2Nb
InChi Key: SZUFSQYBUPXGSX-UHFFFAOYSA-N
SMILES: Cl.Cl.C[c-]1cccc1.C[C-]12[C-]3[C-]4[C-]5[C-]1[Nb]2345
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results