accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3NOHO

CAS RN 6138-41-6

IUPAC Name: 3-carboxy-1-methylpyridin-1-ium chloride
Synonyms: trigonelline
Molecular Weight (g/mol): 173.6
Molecular Formula: C7H8ClNO2
InChi Key: TZSYLWAXZMNUJB-UHFFFAOYSA-N
SMILES: [Cl-].C[N+]1=CC=CC(=C1)C(O)=O
Grade 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)