Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trigonelline Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T14295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14295G Supplier TCI America Supplier No. T14295G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Betaine-type pyrimidine alkaloid with diverse reported biological activities, including hypoglycemic and neuroprotective effects.

Chemical Identifiers

CAS 6138-41-6
Molecular Formula C7H8ClNO2
Molecular Weight (g/mol) 173.60
MDL Number MFCD00077250
InChI Key TZSYLWAXZMNUJB-UHFFFAOYSA-N
Synonym N-Methylnicotinic Acid Betaine Hydrochloride, 1-Methylpyridinium-3-carboxylate Hydrochloride
PubChem CID 134606
IUPAC Name 3-carboxy-1-methylpyridin-1-ium chloride
SMILES [Cl-].C[N+]1=CC=CC(=C1)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 173.60
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Trigonelline Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.