accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6138-90-5

IUPAC Name: (6E)-8-bromo-2,6-dimethylocta-2,6-diene
Synonyms: (6E)-8-bromo-2,6-dimethylocta-2,6-diene
Molecular Weight (g/mol): 217.15
Molecular Formula: C10H17Br
InChi Key: JSCUZAYKVZXKQE-JXMROGBWSA-N
SMILES: CC(C)=CCC\C(C)=C\CBr
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)