Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Geranyl bromide

Catalog No. 3291185G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-911-85G 5 g
32-911-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-911-85G Supplier Sigma Aldrich Supplier No. 3291185G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Geranyl bromide undergoes palladium catalyzed cross-coupling reaction with aryl and alkenylgold(i) phosphanes.geranyl bromide was used in synthesis of baicalein and 3,7-dihydroxyflavone derivatives
  • It was also used in synthesis of potential flavonoidic modulators of p-glycoprotein activity

Specifications

CAS 6138-90-5
Density 1.094 g/mL (at 25°C (literature))
Boiling Point 101°C to 102°C (12 mmHg)
Molecular Formula C10H17Br
Linear Formula (CH3)2 C=CHCH2CH2C(CH3)=CHCH2Br
Refractive Index n20/D 1.504 (literature)
Quantity 5 g
MDL Number MFCD00000243
Synonym trans-1-Bromo-3,7-dimethyl-2,6-octadiene
Molecular Weight (g/mol) 217.15
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.