accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 614-72-2

IUPAC Name: 1-chloro-2-ethoxybenzene
Synonyms: benzene, 1-chloro-2-ethoxy-
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: IRYSAAMKXPLGAM-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results