Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorophenetole 99.0+%, TCI America™
SDP

Catalog No. C023825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C023825ML 25 mL
C023810ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C023825ML Supplier TCI America Supplier No. C023825ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 614-72-2
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00058930
InChI Key IRYSAAMKXPLGAM-UHFFFAOYSA-N
Synonym 1-Chloro-2-ethoxybenzene
PubChem CID 69197
IUPAC Name 1-chloro-2-ethoxybenzene
SMILES CCOC1=CC=CC=C1Cl

Specifications

Color Yellow
Quantity 25 mL
Physical Form Clear Liquid at 20°C
Boiling Point 210°C
Formula Weight 156.61
Percent Purity ≥99.0% (GC)
Chemical Name or Material 2-Chlorophenetole
EINECSNumber (3)-0845
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.