accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 614-94-8

IUPAC Name: 4-methoxybenzene-1,3-diamine dihydrochloride
Synonyms: 2,4 diaminoanisole dihydrochloride
Molecular Weight (g/mol): 211.09
Molecular Formula: C7H12Cl2N2O
InChi Key: FNGHHDCBSVSOOI-UHFFFAOYSA-N
SMILES: Cl.Cl.COC1=C(N)C=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results