Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diaminoanisole Dihydrochloride 99.0+%, TCI America™
SDP

Catalog No. D00745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00745G 5 g
D007425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D00745G Supplier TCI America Supplier No. D00745G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 614-94-8
Molecular Formula C7H12Cl2N2O
Molecular Weight (g/mol) 211.086
MDL Number MFCD00054337
InChI Key FNGHHDCBSVSOOI-UHFFFAOYSA-N
Synonym 4-Methoxy-1,3-phenylenediamine Dihydrochloride
PubChem CID 11975
IUPAC Name 4-methoxybenzene-1,3-diamine;dihydrochloride
SMILES COC1=C(C=C(C=C1)N)N.Cl.Cl

Specifications

Quantity 5 g
Formula Weight 211.09
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diaminoanisole Dihydrochloride
EINECSNumber (3)-0730
RTECSNumber ST2693000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.