accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 6142-95-6

IUPAC Name: (3,3-diethoxyprop-1-yn-1-yl)benzene
Synonyms: (3,3-diethoxyprop-1-yn-1-yl)benzene
Molecular Weight (g/mol): 204.27
Molecular Formula: C13H16O2
InChi Key: DTEGZYXCDQFSBZ-UHFFFAOYSA-N
SMILES: CCOC(OCC)C#CC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results