CAS RN 61444-38-0
IUPAC Name:
(3Z)-hex-3-en-1-yl (3Z)-hex-3-enoate
Synonyms:
cis-3-hexenyl cis-hexenoate
Molecular Weight (g/mol):
196.29
Molecular Formula:
C12H20O2
InChi Key:
UZJQQWFHPLYECS-SFECMWDFSA-N
SMILES:
CC\C=C/CCOC(=O)C\C=C/CC
1
–
1
of
1
results
1
cis-3-Hexenyl cis-3-Hexenoate 95.0+%, TCI America™
CAS: 61444-38-0 Molecular Formula: C12H20O2 Molecular Weight (g/mol): 196.29 MDL Number: MFCD00036652 InChI Key: UZJQQWFHPLYECS-SFECMWDFSA-N Synonym: cis-3-Hexenoic Acid cis-3-Hexenyl Ester PubChem CID: 6435882 IUPAC Name: [(Z)-hex-3-enyl] (Z)-hex-3-enoate SMILES: CCC=CCCOC(=O)CC=CCC