Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Hexenyl cis-3-Hexenoate 95.0+%, TCI America™
SDP

Catalog No. H06775ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06775ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H06775ML Supplier TCI America Supplier No. H06775ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61444-38-0
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00036652
InChI Key UZJQQWFHPLYECS-SFECMWDFSA-N
Synonym cis-3-Hexenoic Acid cis-3-Hexenyl Ester
PubChem CID 6435882
IUPAC Name [(Z)-hex-3-enyl] (Z)-hex-3-enoate
SMILES CCC=CCCOC(=O)CC=CCC

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 196.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3-Hexenyl cis-3-Hexenoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.