accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNH3ONOO

CAS RN 6149-33-3

IUPAC Name: 4-(4-nitrophenoxy)anilinium chloride
Synonyms: 4-(4-nitrophenoxy)anilinium chloride
Molecular Weight (g/mol): 266.68
Molecular Formula: C12H11ClN2O3
InChi Key: FDQAGKDMDFYNBL-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]C1=CC=C(OC2=CC=C(C=C2)[N+]([O-])=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results