Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-4'-nitrodiphenyl Ether 98.0+%, TCI America™
SDP

Catalog No. A191025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A191025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A191025G Supplier TCI America Supplier No. A191025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6149-33-3
Molecular Formula C12H10N2O3
Molecular Weight (g/mol) 230.223
InChI Key ASAOLTVUTGZJST-UHFFFAOYSA-N
PubChem CID 80251
IUPAC Name 4-(4-nitrophenoxy)aniline
SMILES C1=CC(=CC=C1N)OC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 135°C
Quantity 25 g
Formula Weight 230.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-4′-nitrodiphenyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.