CAS RN 615-66-7
IUPAC Name:
2-chlorobenzene-1,4-diamine; sulfuric acid
Synonyms:
2-chloro-P-phenylenediamine
Molecular Weight (g/mol):
240.66
Molecular Formula:
C6H9ClN2O4S
InChi Key:
GQFGHCRXPLROOF-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.NC1=CC=C(N)C(Cl)=C1
1
–
1
of
1
results
1
2-Chloro-1,4-phenylenediamine 98.0+%, TCI America™
CAS: 615-66-7 Molecular Formula: C6H7ClN2 Molecular Weight (g/mol): 142.586 MDL Number: MFCD00128931 InChI Key: MGLZGLAFFOMWPB-UHFFFAOYSA-N Synonym: 1,4-Diamino-2-chlorobenzene PubChem CID: 11998 ChEBI: CHEBI:76598 IUPAC Name: 2-chlorobenzene-1,4-diamine SMILES: C1=CC(=C(C=C1N)Cl)N