Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,4-phenylenediamine 98.0+%, TCI America™
SDP

Catalog No. C305425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C305425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C305425G Supplier TCI America Supplier No. C305425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 615-66-7
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD00128931
InChI Key MGLZGLAFFOMWPB-UHFFFAOYSA-N
Synonym 1,4-Diamino-2-chlorobenzene
PubChem CID 11998
ChEBI CHEBI:76598
IUPAC Name 2-chlorobenzene-1,4-diamine
SMILES C1=CC(=C(C=C1N)Cl)N

Specifications

Melting Point 65°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 142.59
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2-Chloro-1,4-phenylenediamine
EINECSNumber (3)-0267
RTECSNumber SS8925000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.