accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOONHOHN

CAS RN 6153-44-2

IUPAC Name: methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Synonyms: methyl 2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate
Molecular Weight (g/mol): 170.12
Molecular Formula: C6H6N2O4
InChi Key: UUTDWTOZAWFKFW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=O)NC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results