accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH(R)NH2OOHClCH3

CAS RN 61535-49-7

IUPAC Name: methyl (2R)-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
Synonyms: methyl (2R)-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
Molecular Weight (g/mol): 254.71
Molecular Formula: C12H15ClN2O2
InChi Key: XNFNGGQRDXFYMM-HNCPQSOCSA-N
SMILES: Cl.COC(=O)[C@H](N)CC1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)