accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61597-98-6

IUPAC Name: 5-methyl-2-(propan-2-yl)cyclohexyl 2-hydroxypropanoate
Synonyms: menthyl lactate
Molecular Weight (g/mol): 228.33
Molecular Formula: C13H24O3
InChi Key: UJNOLBSYLSYIBM-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C)CC1OC(=O)C(C)O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results