Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl (S)-2-Hydroxypropionate 98.0+%, TCI America™
SDP

Catalog No. I08895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08895G 5 g
I088925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08895G Supplier TCI America Supplier No. I08895G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61597-98-6
Molecular Formula C13H24O3
Molecular Weight (g/mol) 228.33
MDL Number MFCD09037384,MFCD27977194,MFCD00589639
InChI Key UJNOLBSYLSYIBM-UHFFFAOYNA-N
Synonym (S)-2-Hydroxypropionic Acid (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl Ester, L-Menthyl L-Lactate
PubChem CID 7076215
IUPAC Name 5-methyl-2-(propan-2-yl)cyclohexyl 2-hydroxypropanoate
SMILES CC(C)C1CCC(C)CC1OC(=O)C(C)O

Specifications

Melting Point 45°C
Color White
Quantity 5 g
Formula Weight 228.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl (S)-2-Hydroxypropionate
EINECSNumber (3)-3279
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.