accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 616-02-4

IUPAC Name: 3-methyl-2,5-dihydrofuran-2,5-dione
Synonyms: 2,5-furandione, 3-methyl-
Molecular Weight (g/mol): 112.08
Molecular Formula: C5H4O3
InChi Key: AYKYXWQEBUNJCN-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC1=O
Boiling Point 
  • (4)
  • (6)
  • (3)
Molecular Weight (g/mol) 
  • (7)
  • (6)
Percent Purity 
  • (3)
  • (10)
Quantity 
  • (1)
  • (4)
  • (4)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (3)
  • (7)
  • (4)
  • (6)
  • (3)
  • (7)
  • (6)
  • (3)
  • (10)
  • (1)
  • (4)
  • (4)
  • (1)
  • (3)
  • (7)