accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 616-04-6

CH3NOHNO

CAS RN 616-04-6

IUPAC Name: 1-methylimidazolidine-2,4-dione
Synonyms: 1-methylhydantoin dioxy-creatinine
Molecular Weight (g/mol): 114.1
Molecular Formula: C4H6N2O2
InChi Key: RHYBFKMFHLPQPH-UHFFFAOYSA-N
SMILES: CN1CC(=O)NC1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results