Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylhydantoin 99.0+%, TCI America™
SDP

Catalog No. M102725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M102725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M102725G Supplier TCI America Supplier No. M102725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 616-04-6
Melting Point 158°C
Color White
Molecular Formula C4H6N2O2
Quantity 25 g
MDL Number MFCD00003187
Synonym 1-methylhydantoin, dioxy-creatinine, n-methylhydantoin, hydantoin, 1-methyl, 1-methyl-hydantoin, methylhydantoin, n-methylimidazolidine-2,4-dione, unii-0546h9ur1r, 1-methyldiazolidine-2,4-dione, 2,4-imidazolidinedione, 1-methyl
InChI Key RHYBFKMFHLPQPH-UHFFFAOYSA-N
SMILES CN1CC(=O)NC1=O
IUPAC Name 1-methylimidazolidine-2,4-dione
Molecular Weight (g/mol) 114.104
PubChem CID 69217
ChEBI CHEBI:16354
Formula Weight 114.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylhydantoin
Show More Show Less
EINECSNumber (5)-6017
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.