accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 616-09-1

H3COOCH3OH

CAS RN 616-09-1

IUPAC Name: propyl 2-hydroxypropanoate
Synonyms: propyl 2-hydroxypropanoate
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: ILVGAIQLOCKNQA-UHFFFAOYNA-N
SMILES: CCCOC(=O)C(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results