Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl Lactate 95.0+%, TCI America™
SDP

Catalog No. L012125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L012125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L012125G Supplier TCI America Supplier No. L012125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 616-09-1
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00059661
InChI Key ILVGAIQLOCKNQA-UHFFFAOYSA-N
Synonym Lactic Acid Propyl Ester
PubChem CID 92821
IUPAC Name propyl 2-hydroxypropanoate
SMILES CCCOC(=O)C(C)O

Specifications

Color Colorless
Boiling Point 169°C
Quantity 25 g
Formula Weight 132.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Lactate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.