accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 616-13-7

IUPAC Name: 1-chloro-2-methylbutane
Synonyms: 1-chloro-2-methylbutane
Molecular Weight (g/mol): 106.59
Molecular Formula: C5H11Cl
InChi Key: IWAKWOFEHSYKSI-UHFFFAOYNA-N
SMILES: CCC(C)CCl
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)