Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2-methylbutane 95.0+%, TCI America™
SDP

Catalog No. C14595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C14595G Supplier TCI America Supplier No. C14595G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 616-13-7
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.593
MDL Number MFCD00039366
InChI Key IWAKWOFEHSYKSI-UHFFFAOYSA-N
PubChem CID 12015
IUPAC Name 1-chloro-2-methylbutane
SMILES CCC(C)CCl

Specifications

Color Yellow
Boiling Point 99°C
Quantity 5 g
UN Number 1107
Formula Weight 106.59
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-methylbutane
EINECSNumber (2)-0066
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.